Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646751.1 | XP_007644941.1 | 4 - 5 | 501 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Probe spans exons |