Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653647.1 | XP_007651837.1 | 5 - 6 | 818 | 105 | |
GenBank mRNA | JI864476.1 | - | 5 - 6 | 825 | 105 | |
JP045701.1 | - | 5 - 6 | 825 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |