Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513331.2 | XP_003513379.1 | 3 | 1144 | 99 | |
XM_007653630.1 | XP_007651820.1 | 2 | 415 | 99 | ||
XM_007653631.1 | XP_007651821.1 | 2 | 309 | 99 | ||
GenBank mRNA | JI885160.1 | - | 2 | 338 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |