Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699135.3 | XP_005699192.1 | 4 - 5 | 1402 | 59 | |
GenBank mRNA | JO590903.2 | - | 2 - 3 | 168 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Capra hircus Probe spans exons |