Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059511.1 | XP_017915000.1 | 4 - 5 | 991 | 66 | |
XM_018059512.1 | XP_017915001.1 | 5 - 6 | 586 | 66 | ||
XM_018059513.1 | XP_017915002.1 | 3 - 4 | 348 | 66 | ||
GenBank mRNA | JO369533.2 | - | 1 - 2 | 378 | 66 | |
JO577012.1 | - | 2 - 3 | 801 | 66 | ||
JO577013.1 | - | 2 - 3 | 801 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |