Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053687.1 | XP_017909176.1 | 2 - 3 | 264 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |