Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689567.3 | XP_005689624.1 | 3 - 4 | 652 | 52 | |
XM_005689568.3 | XP_005689625.1 | 2 - 3 | 1055 | 52 | ||
XM_018059625.1 | XP_017915114.1 | 2 - 3 | 1055 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |