Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695172.3 | XP_005695229.2 | 2 - 3 | 461 | 71 | |
GenBank mRNA | JO446150.2 | - | 1 - 2 | 387 | 71 | |
KA306556.1 | - | 1 - 2 | 452 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |