Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051717.1 | XP_017907206.1 | 3 - 4 | 1021 | 77 | |
XM_018051718.1 | XP_017907207.1 | 1 - 2 | 1314 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |