Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042127.1 | XP_017897616.1 | 3 - 4 | 948 | 85 | |
XM_018042128.1 | XP_017897617.1 | 3 - 4 | 780 | 85 | ||
GenBank mRNA | JP350800.2 | - | 1 - 2 | 831 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |