Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677737.3 | XP_005677794.1 | 1 - 2 | 118 | 79 | |
GenBank mRNA | JO387383.2 | - | 3 - 4 | 402 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |