Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678989.3 | XP_005679046.2 | 2 - 3 | 982 | 80 | |
GenBank mRNA | JT756169.1 | - | 2 - 3 | 1294 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |