Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052558.1 | XP_017908047.1 | 3 - 4 | 1955 | 85 | |
XM_018052559.1 | XP_017908048.1 | 3 - 4 | 1847 | 85 | ||
XM_018052560.1 | XP_017908049.1 | 4 - 5 | 1856 | 85 | ||
GenBank mRNA | KA329193.1 | - | 962 | 85 | ||
KU041754.1 | - | 4 - 5 | 1337 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |