Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBEGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050289.1 | XP_017905778.1 | 6 - 7 | 1470 | 95 | |
XM_018050290.1 | XP_017905779.1 | 6 - 7 | 1867 | 95 | ||
GenBank mRNA | KA336377.1 | - | 5 - 6 | 950 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |