Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059234.1 | XP_017914723.1 | 4 - 5 | 512 | 66 | |
XM_018059235.1 | XP_017914724.1 | 4 - 5 | 509 | 66 | ||
XM_018059236.1 | XP_017914725.1 | 4 - 5 | 560 | 66 | ||
GenBank mRNA | JO426523.1 | - | 4 - 5 | 685 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |