Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681841.3 | XP_005681898.2 | 3 - 4 | 725 | 91 | |
XM_005681842.3 | XP_005681899.2 | 3 - 4 | 626 | 91 | ||
XM_013964756.2 | XP_013820210.2 | 3 - 4 | 717 | 91 | ||
GenBank mRNA | JO594467.2 | - | 3 - 4 | 712 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |