Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARTPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694591.3 | XP_005694648.1 | 1 - 2 | 440 | 92 | |
GenBank mRNA | JO625361.2 | - | 1 - 2 | 201 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |