Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285666.1 | NP_001272595.1 | 2 - 3 | 249 | 98 | |
XM_005677471.3 | XP_005677528.1 | 2 - 3 | 478 | 98 | ||
XM_005677473.3 | XP_005677530.1 | 2 - 3 | 293 | 98 | ||
GenBank mRNA | HQ322407.1 | - | 2 - 3 | 249 | 98 | |
JT686393.1 | - | 2 - 3 | 298 | 98 | ||
KA305224.1 | - | 2 - 3 | 1776 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |