Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285567.1 | NP_001272496.1 | 2 - 3 | 142 | 67 | |
GenBank mRNA | JF731041.1 | - | 2 - 3 | 142 | 67 | |
JO580392.1 | - | 2 - 3 | 312 | 67 | ||
KA334415.1 | - | 2 - 3 | 294 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |