Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285733.1 | NP_001272662.1 | 1 - 2 | 502 | 79 | |
GenBank mRNA | JF829006.1 | - | 1 - 2 | 502 | 79 | |
KX056119.2 | - | 1 - 2 | 505 | 79 | ||
KX171206.1 | - | 1 - 2 | 505 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |