Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285759.1 | NP_001272688.1 | 3 - 4 | 378 | 82 | |
GenBank mRNA | AY389343.1 | - | 3 - 4 | 378 | 82 | |
EF199763.1 | - | 3 - 4 | 341 | 82 | ||
GQ369727.1 | - | 3 - 4 | 329 | 82 | ||
JN377801.1 | - | 2 - 3 | 284 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |