Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314285.1 | NP_001301214.1 | 2 - 3 | 165 | 110 | |
XM_005690078.3 | XP_005690135.1 | 2 - 3 | 527 | 110 | ||
XM_005690080.3 | XP_005690137.1 | 2 - 3 | 254 | 110 | ||
GenBank mRNA | JT698247.1 | - | 10 - 11 | 1705 | 110 | |
KA331617.1 | - | 5 - 6 | 725 | 110 | ||
KF669900.1 | - | 2 - 3 | 165 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |