Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692684.3 | XP_005692741.2 | 6 - 7 | 1384 | 72 | |
XM_018062726.1 | XP_017918215.1 | 6 - 7 | 1389 | 72 | ||
XM_018062727.1 | XP_017918216.1 | 5 - 6 | 1197 | 72 | ||
GenBank mRNA | JO633226.2 | - | 6 - 7 | 1325 | 72 | |
JO633228.2 | - | 6 - 7 | 1325 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |