Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684085.3 | XP_005684142.1 | 4 - 5 | 472 | 109 | |
XM_005684087.3 | XP_005684144.1 | 4 - 5 | 470 | 109 | ||
XM_013966145.2 | XP_013821599.1 | 4 - 5 | 494 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |