Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038729.1 | XP_017894218.1 | 2 - 3 | 1572 | 89 | |
XM_018038730.1 | XP_017894219.1 | 2 - 3 | 1572 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |