Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040209.1 | XP_017895698.1 | 2 - 3 | 217 | 83 | |
GenBank mRNA | JO580117.1 | - | 2 - 3 | 214 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |