Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052251.1 | XP_017907740.1 | 3 - 4 | 1516 | 89 | |
XM_018052253.1 | XP_017907742.1 | 4 - 5 | 1619 | 89 | ||
GenBank mRNA | JO648091.2 | - | 1 - 2 | 109 | 89 | |
KA312111.1 | - | 1 - 2 | 1825 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |