Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO628644.2 | - | 4 - 5 | 805 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |