Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691881.3 | XP_005691938.3 | 4 - 5 | 1014 | 69 | |
GenBank mRNA | JO364249.2 | - | 4 - 5 | 1236 | 69 | |
KA325250.1 | - | 4 - 5 | 907 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |