Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695348.3 | XP_005695405.1 | 3 - 4 | 1126 | 100 | |
XM_005695349.3 | XP_005695406.1 | 3 - 4 | 1106 | 100 | ||
GenBank mRNA | JO420012.2 | - | 3 - 4 | 1127 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |