Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEPACAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043577.1 | XP_017899066.1 | 4 - 5 | 1184 | 95 | |
XM_018043578.1 | XP_017899067.1 | 4 - 5 | 1184 | 95 | ||
GenBank mRNA | JO632295.2 | - | 4 - 5 | 1216 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |