Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HENMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678010.3 | XP_005678067.2 | 4 - 5 | 422 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |