Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H9orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687300.3 | XP_005687357.2 | 7 - 8 | 754 | 84 | |
XM_018055961.1 | XP_017911450.1 | 6 - 7 | 683 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |