Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042601.1 | XP_017898090.1 | 5 - 6 | 1500 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |