Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064012.1 | XP_017919501.1 | 11 - 12 | 1032 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |