Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699878.3 | XP_005699935.1 | 3 - 4 | 409 | 64 | |
XM_018043488.1 | XP_017898977.1 | 3 - 4 | 411 | 64 | ||
XM_018043489.1 | XP_017898978.1 | 3 - 4 | 351 | 64 | ||
XM_018043490.1 | XP_017898979.1 | 3 - 4 | 409 | 64 | ||
GenBank mRNA | JI861828.1 | - | 2 - 3 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |