Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049079.1 | XP_017904568.1 | 12 - 13 | 2288 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |