Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690159.3 | XP_005690216.1 | 7 - 8 | 892 | 63 | |
XM_005690160.3 | XP_005690217.1 | 7 - 8 | 896 | 63 | ||
GenBank mRNA | JO630283.2 | - | 2 - 3 | 436 | 63 | |
JT698315.1 | - | 9 - 10 | 821 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |