Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059818.1 | XP_017915307.1 | 4 - 5 | 1292 | 107 | |
XM_018059819.1 | XP_017915308.1 | 4 - 5 | 1187 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |