Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6H4orf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049278.1 | XP_017904767.1 | 13 - 14 | 1540 | 100 | |
XM_018049279.1 | XP_017904768.1 | 12 - 13 | 1505 | 100 | ||
XM_018049280.1 | XP_017904769.1 | 12 - 13 | 1463 | 100 | ||
XM_018049281.1 | XP_017904770.1 | 12 - 13 | 1210 | 100 | ||
XM_018049282.1 | XP_017904771.1 | 12 - 13 | 1340 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |