Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059046.1 | XP_017914535.1 | 1 - 2 | 418 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |