Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969058.2 | XP_013824512.2 | 3 - 4 | 357 | 79 | |
XM_018058667.1 | XP_017914156.1 | 4 - 5 | 600 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |