Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066203.1 | XP_017921692.1 | 4 - 5 | 1202 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |