Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038371.1 | XP_017893860.1 | 2 - 3 | 132 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |