Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681883.3 | XP_005681940.2 | 3 - 4 | 232 | 94 | |
GenBank mRNA | JO623459.2 | - | 3 - 4 | 1545 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |