Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698317.3 | XP_005698374.2 | 3 - 4 | 700 | 81 | |
GenBank mRNA | JO622213.2 | - | 1 - 2 | 850 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |