Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044744.1 | XP_017900233.1 | 3 - 4 | 754 | 74 | |
GenBank mRNA | JO581425.1 | - | 3 - 4 | 711 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |