Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054929.1 | XP_017910418.1 | 2 - 3 | 782 | 67 | |
GenBank mRNA | JO618519.2 | - | 3 - 4 | 842 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |