Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684480.2 | XP_005684537.1 | 1 - 2 | 323 | 98 | |
GenBank mRNA | JO584837.2 | - | 2 - 3 | 412 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |