Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700487.2 | XP_005700544.1 | 6 - 7 | 1012 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |